Learn More
Medchemexpress LLC 4H-1-Benzopyran-4-one, 8-β-D-glucopyranosyl-7-hydroxy-3-(4-methoxyphenyl)- | 92117-94-7 | 430.40 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN19791MG
Encompass_Preferred
This product is 4H-1-Benzopyran-4-one, 8-β-D-glucopyranosyl-7-hydroxy-3-(4-methoxyphenyl)-, presented as a white to off-white solid with a purity of 99.70% by HPLC. It is intended for research use only and has not been fully validated for medical applications.
  • White to off-white solid appearance
  • 99.70% purity (HPLC)
  • Sealed storage at 4°C, away from moisture and light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month (sealed storage, away from moisture and light)
  • Complies with given specifications
Catalog No. 50-004-14614