Learn More
Medchemexpress LLC 4H-Pyrazolo[3,4-d]pyrimidin-4-one, 1,5-dihydro-6-[(1S)-1-(3-phenoxy-1-azetidinyl)ethyl]-1-(tetrahydro-2H-pyran-4-yl)- | 99.9% | 25 MG

Supplier: Medchemexpress LLC 25MGHY12788A
(S)-PF-04449613 is the left-handed isomer of PF-04449613, a selective PDE9A inhibitor. It exhibits an IC50 of 22 nM and has been observed to improve motor learning ability in a mouse model. This compound is provided as a white to light yellow solid.
- Purity of 99.9%
- Selective PDE9A inhibitor (IC50: 22 nM)
- Improves motor learning ability in mouse models
- Molecular weight of 395.45
- Molecular formula: C21H25N5O3
- Soluble in DMSO: 100 mg/mL
- Stable for 3 years as powder at -20°C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.