Learn More
Medchemexpress LLC 4H-Pyrimido[6,1-a]isoquinolin-4-one, 2,3,6,7-tetrahydro-9,10-dimethoxy-3-methyl-2-[(2,4,6-trimethylphenyl)imino]-, hydrochloride (1:1) | 78416-81-6 | 99.86% | 441.95 | 10 MM 1 ML
SDP
Supplier:  Medchemexpress LLC HY18740A10MM/1ML
Encompass_Preferred
Trequinsin hydrochloride (HL 725) is an extremely potent inhibitor of platelet CAMP phosphodiesterase (PDE), with an IC50 of 0.25 nM. It is also an extremely potent inhibitor of the aggregation of human platelets induced in vitro by ADP, collagen, thrombin, and epinephrine.
  • Extremely potent inhibitor of platelet CAMP phosphodiesterase (PDE)
  • IC50 of 0.25 nM for PDE inhibition
  • Potent inhibitor of human platelet aggregation induced by ADP, collagen, thrombin, and epinephrine
  • Exerts cardiovascular and antihypertensive qualities
  • Efficacious agonist of [Ca2+]i
Catalog No. 50-003-85719