Learn More
Cayman Chemical 4hydroxy Diclofenac
SDP
Supplier:  Cayman Chemical 100085181 MG
Encompass_Preferred
An active metabolite of diclofenac; inhibits COX activity (IC50 = 32 nM) and reduces PGE2 production (IC50 = 17 nM) in isolated human rheumatoid synovial cells
Catalog No. 50-288-7673