Learn More
Cayman Chemical 4iodoSAHA Inhibitors
SDP
Supplier:  Cayman Chemical 10495250 MG
Encompass_Preferred
A hydrophobic derivative of the class I and class II HDAC inhibitor SAHA that demonstrates >60% inhibition of HDAC1 and HDAC6 activity in a deacetylase activity assay; inhibits proliferation of SK-BR-3 breast-derived, HT29 colon-derived, and U937 leukemia cell lines with EC50 values of 1.1, 0.95, and 0.12 μM, respectively
Catalog No. 50-289-1070