Learn More
Cayman Chemical 4methyl Acetyl fentnyld5 100ug
SDP
Supplier:  Cayman Chemical 35276100 UG
Encompass_Preferred
An internal standard for the quantification of 4’-methyl acetyl fentanyl by GC- or LC-MS; regulated as a Schedule I compound in the United States; intended for research and forensic applications
Catalog No. 50-290-9545