Learn More
Cayman Chemical 4Methyl7 4nItro2 trIflu 10mg
SDP
Supplier:  Cayman Chemical 2847110 MG
Encompass_Preferred
A turn-on fluorescent probe for H2S; excited at 348 nm in DMSO and fluorescence intensity increases in the presence of HS-; selective for HS- over cysteine, AcO-, F-, H2PO4-, Cl-, Br-, and I- and is not cytotoxic up to a concentration of 150 UG/ml
Catalog No. 50-293-7816