Learn More
Cayman Chemical 4MethylumbelIferylDGluc 500mg
SDP
Supplier:  Cayman Chemical 20948500 MG
Encompass_Preferred
A fluorogenic substrate of β-glucosidase and β-glucocerebrosidase
Catalog No. 50-298-3241