Learn More
Cayman Chemical 4MethylumbelIferylDGluc 500mg
SDP
Supplier:  Cayman Chemical 17203500 MG
Encompass_Preferred
A fluorogenic substrate of β-glucuronidase; cleaved by β-glucuronide to release the fluorescent moiety 4-MU
Catalog No. 50-296-6237