Learn More
Chemglass Life Sciences Nest Scientific 4 mL Sterile Transport Tubes with Caps on, 50/pack, 1,000/case
SDP
Supplier:  Chemglass Life Sciences CGN474204
Encompass_Preferred
Nest Scientific 4 mL sterile transport tubes with caps on are disposable polypropylene sample tubes designed for secure collection, transport, and short-term storage of biological specimens. Manufactured in a controlled clean-room environment and electron beam sterilized to SAL=10^-6, the tubes are DNase-, RNase-, pyrogen-, and endotoxin-free, and the HDPE caps provide a leak-resistant seal for safe air transport.
  • 4 mL capacity.
  • USP VI polypropylene tube with HDPE cap.
  • Sterile; electron beam sterilized to SAL=10^-6.
  • Packaged 50 per pack, 1,000 per case.
  • Temperature range approximately -196°C to 121°C.
  • DNase-, RNase-, pyrogen-, and endotoxin-free.
  • Self-standing design for easy handling and storage.
  • Leak-resistant sealing; tested to negative pressure 95 kPa for air transport.
Catalog No. 50-273-3406