Learn More
Cayman Chemical 4oxo DocosahexaenoIc ACD 100ug
SDP
Supplier:  Cayman Chemical 21373100 UG
Encompass_Preferred
An active metabolite of docosahexaenoic acid; agonist of PPARγ; induces gene transcription in a reporter assay using COS-1 cells expressing PPARγ (EC50 = ~10 µM); decreases the proliferation of MDA-MB-231, BT-549, T47D, and SK-BR-3 cancer cells, MCF-10F cells, and increases the proliferation of MCF-7 cancer cells at 100 µM
Catalog No. 50-298-5536