Learn More
Cayman Chemical 4oxo27TBDMS WIthaferIn A 10mg
SDP
Supplier:  Cayman Chemical 2832310 MG
Encompass_Preferred
A derivative of withaferin A; is cytotoxic to A2780 ovarian cancer cells (IC50 = 17 µM); selective for A2780 cells over non-cancerous ARPE19 cells (IC50 = 1,660 µM); induces DNA fragmentation in A2780 cells
Catalog No. 50-294-1533