Learn More
Cayman Chemical 4oxo2Nonenal Fatty Acids and D
SDP
Supplier:  Cayman Chemical 101851 MG
Encompass_Preferred
A lipid peroxidation product; actively modifies histidine and lysine residues on proteins and causes protein cross-linking
Catalog No. 50-288-8407