Learn More
Andwin Scientific 4QT SQUARE FOOD CONTAINER 6CS
SDP
Supplier:  Andwin Scientific 4SFSPROCW135 KTM
Encompass
Part of the CamSquare FreshPro series4-qt. capacityClear Camwear polycarbonateBreak, stain, acid, and odor resistantConcaved, textured gripRecessed handles with drain holes for dryingCurved corners enable four-sided pouringGreen, color-coded graduationsTemperature range: - 40 to 210 degrees FStackable ledgeDishwasher safeOverall dimensions: 71⁄2 in. L x 71⁄2 in. W x 71/5 in. H
Catalog No. NC3314505