Learn More
Cayman Chemical 4QuInoLN3CarboxamIde Fur 10mg
SDP
Supplier:  Cayman Chemical 1109410 MG
Encompass_Preferred
A high-affinity ligand of CB2 (Ki = 8.5 nM) with little affinity for CB1 (Ki >10,000 nM); demonstrates antinociceptive efficacy in the mouse formalin test at 1 mg/kg, an action which is blocked by a CB2-selective antagonist, AM630
Catalog No. 50-295-1251