Learn More
Medchemexpress LLC (4R)-1-{[4,4-difluoro-1-(4-methoxyphenyl)cyclohexyl]carbonyl}-4-fluoro-N-1H-pyrazolo[4,3-b]pyridin-5-yl-d-prol... | 2259641-46-6 | 99.8% | 501.5 g·mol-1 | C25H26F3N5O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15224110MG
Encompass_Preferred
DS-9300 is a small-molecule, orally active, selective inhibitor of the EP300/CBP histone acetyltransferase (HAT) with a reported biochemical IC50 of 28 nM. Described in the Journal of Medicinal Chemistry and developed by Daiichi Sankyo, the compound has shown anticancer activity in preclinical prostate cancer models.
  • Potent EP300/CBP HAT inhibition (IC50 = 28 nM).
  • Orally active with demonstrated antitumor effects in preclinical prostate cancer models.
  • Selective activity for EP300/CBP over other HATs reported in the discovery study.
  • Provided as small-scale research quantities with analytical documentation available.
  • High reported purity (99.83%) and molecular formula C25H26F3N5O3.
  • Molecular weight approximately 501.5 g·mol-1.
Catalog No. 50-002-86410