Learn More
Medchemexpress LLC (4R)-5-(dipentylamino)-5-oxo-4-(quinoline-3-carbonylamino)pentanoic acid | 119295-94-2 | 99.9% | 441.56 g/mol | C25H35N3O4 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1181941MG
Encompass_Preferred
A selective cholecystokinin A (CCK-A) receptor antagonist used in research to inhibit CCK8-induced amylase secretion. Supplied as a high-purity, white to off-white solid with recommended cold storage to maintain stability.
  • Selective CCK-A receptor antagonist for receptor pharmacology studies.
  • High purity suitable for biochemical and cellular assays.
  • White to off-white solid appearance for easy visual inspection.
  • Stable under recommended cold storage for extended shelf life.
  • Compatible with common in vitro solvent systems when handled as directed.
Catalog No. 50-003-07838