Learn More
Strem, An Ascensus Company CAS# 32305-98-9. 250mg. (4R,5R)-(-)-4,5-Bis(diphenylphosphinomethyl)-2,2-dimethyl-1,3-dioxolane, 99.5% (R,R)-DIOP. MFCD00014107
SDP
Supplier:  Strem, An Ascensus Company 152970250MG
Encompass
CAS# 32305-98-9. 250mg. (4R,5R)-(-)-4,5-Bis(diphenylphosphinomethyl)-2,2-dimethyl-1,3-dioxolane, 99.5% (R,R)-DIOP. MFCD00014107. Molecular Weight: 498.55. Molecular Formula: C31H32O2P2. Color/form: white pwdr. Strem# 15-2970. http://www.strem.com/catalog/v/15-2970/
Catalog No. 50-901-10246