Learn More
Medchemexpress LLC (4S)-2-oxo-1,4,7,10-tetraazacyclododecane-1,4,7-triacetic acid anhydride | 1375475-53-8 | MFCD33403099 | 99.8% | 458.46 | C19H30N4O9 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW087187100MG
Encompass_Preferred
DOTAGA-anhydride is a DOTA-based bifunctional metal chelator used to bind radionuclides and prepare radionuclide drug conjugates for imaging and therapeutic research. It is supplied in solid and solution forms and is suitable for conjugation to biomolecules for SPECT, PET, or optical imaging applications.
  • DOTA-based bifunctional metal chelator.
  • Enables radiolabeling for SPECT and PET imaging.
  • Suitable for conjugation to biomolecules such as antibodies.
  • Available in solid and solution forms for flexibility in protocols.
  • High purity (approximately 99.8%), suitable for research use.
Catalog No. 50-002-08531