Learn More
Medchemexpress LLC 4SC-203 | 895533-09-2 | 98.98% | 642.77 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY19897100MG
Encompass_Preferred
4SC-203 is a potent multikinase inhibitor with potential antineoplastic activity. It is suitable for research use only.
  • Selectively targets FLT3/STK1
  • Targets FLT3 mutated forms
  • Targets VEGFRs
Catalog No. 50-004-01632