Learn More
Uline Desktop Weatherproof Thermal Transfer Labels - Polypropylene | White | 500
Supplier:  Uline S22434CS
Encompass_Preferred
Smudge proof for plant nurseries, lumberyards and shipping, Matte finish, UV and moisture resistant Use outdoors for up to 1 year, Use with Wax/Resin Ribbons, Perforated between each label
Catalog No. NC2754852