Learn More
GNOSKO BIO INC 4x5 Clear Polymer Freezer Rack
Supplier:  GNOSKO BIO INC GB00CV2
Encompass_Preferred
ChillView freezer racks address common issues with stainless steel freezer boxes. ChillView racks are clear and don't frost, so they don't require gloves to handle. Everything can be seen when opening the door, greatly reducing the time and energy wasted when searching for something specific. Energy is also greatly reduced since ChillView racks transfer heat at approx. 100x less than the rate of stainless-steel boxes. ChillView also allows RFID tags to be read through the box to enable Gnosko Bio automated inventory management solutions. 4 shelves, 5 boxes deep.
Catalog No. 50-312-6228