Learn More
Medchemexpress LLC (4Z)-cyclooct-4-en-1-ol | 85081-69-2 | MFCD06252442 | 99.9% | C8H14O | 10MG
SDP
Supplier:  Medchemexpress LLC HY14118610MG
Encompass_Preferred
TCO-OH (trans-cyclooctene alcohol) is a trans-cyclooctene derivative bearing a hydroxyl group used as a PROTAC linker and click-chemistry handle. It is supplied as a research reagent for synthetic chemistry applications, enabling tetrazine ligation, bioconjugation, and assembly of drug-linker constructs.
  • Facilitates rapid tetrazine ligation for bioorthogonal conjugation.
  • Provides a reactive hydroxyl handle for linker functionalization.
  • High reported purity suitable for analytical and preparative work.
  • Available in small milligram to gram scales for research use.
  • Stable under recommended low-temperature storage conditions.
Catalog No. 50-002-60899