Learn More
Sklar Instruments Single Use, Econo-Sterile, Kelly Hemostatic Forceps, Sklar 5-1/2 in., Blunt, Curved, Serrated, Stainless Steel, Disposable, CS of 25.
SDP
Supplier:  Sklar Instruments 962563
Encompass_Preferred
Single Use, Econo-Sterile, Kelly Hemostatic Forceps, Sklar, 5-1/2 in., Blunt, Curved, Serrated, Stainless Steel, Disposable, CS of 25. Kelly Hemostatic Forceps are primarily used for clamping large blood vessels or manipulating heavy tissue.
Catalog No. 50-109-4277