Learn More
Medchemexpress LLC 5,10,15,20-tetrakis (4-sulfonatophenyl) porphyrin iron III chloride | 90384-82-0 | 99.0% | C44H28ClFeN4O12S4 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13169710MG
Encompass_Preferred
FeTPPS (5,10,15,20-tetrakis(4-sulfonatophenyl)porphyrin iron(III) chloride) is an iron porphyrin reagent used in biochemical research as a peroxynitrite scavenger and decomposition catalyst. It is provided as a high-purity compound for in vitro studies, nitration inhibition experiments, and mechanistic investigations of reactive nitrogen species.
  • High purity (99.0%) suitable for research use.
  • Functions as a peroxynitrite scavenger and anti-nitrating agent.
  • Aids studies of reactive nitrogen species and oxidative stress pathways.
  • Water-soluble sulfonated porphyrin scaffold for biochemical assays.
  • Available in multiple small-mass package sizes for lab-scale experiments.
Catalog No. 50-002-60587