Learn More
Medchemexpress LLC 5-((14-Aminotetradecyl)oxy)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione hydrochloride | 97.7% | 522.08 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1495715MG
Encompass_Preferred
Thalidomide-5-O-C14-NH2 hydrochloride is a Thalidomide (HY-10984)-based cereblon ligand. It is utilized in the recruitment of CRBN protein and can be connected to a ligand for protein by a linker to form PROTACs. This compound is for research use only.
  • Utilized in the recruitment of CRBN protein
  • Can be connected to a ligand for protein by a linker
  • Forms PROTACs
Catalog No. 50-003-61034