Learn More
Chemglass Life Sciences Chemglass Ground Rod, 5/16 in OD x 12 in, 303 Stainless Steel - For Connecting Velp Overhead Stirrers to CG-2047 Flexible Drive Shaft
SDP
Supplier:  Chemglass Life Sciences CG203445
Encompass_Preferred
This 5/16 in OD x 12 in ground rod is machined from 303 stainless steel to provide a durable, corrosion-resistant connection between an overhead stirrer and a flexible drive shaft used in laboratory stirring setups.
  • Durable 303 stainless steel construction for corrosion resistance.
  • 5/16 in outside diameter and 12 in length for standard shaft connections.
  • Provides a secure mechanical connection between overhead stirrers and flexible drive shafts.
  • Sold as an individual item for replacement or accessory use.
Catalog No. NC2326979