Learn More
Ace Glass, Inc. PTFE 5/16in tube compression fitting to 1/8in male NPT
SDP
Supplier:  Ace Glass, Inc. 1270910
Encompass
5/16 PTFE tube compression fitting to 1/8 male NPT. All wetted surfaces are virgin PTFE. Nuts and gripper are PVDF.
Catalog No. 50-977-671