Learn More
Medchemexpress LLC 5-(2,5-dimethylphenoxy)-2,2-dimethylpentanoic acid | 25812-30-0 | MFCD00079335 | 250.33 g/mol | C15H22O3 | 10 MG

Supplier: Medchemexpress LLC HYB0258R10MG
Gemfibrozil analytical standard supplied as a small powder intended for research and analytical use. It serves as a reference standard for assay development and quantitation; chemically, it is a PPAR-α activator and a nonselective inhibitor of several cytochrome P450 isoforms.
- Analytical standard for calibration and quantification.
- Known activator of PPAR-α and nonselective P450 inhibitor.
- Chemical formula C15H22O3; molecular weight 250.33 g/mol.
- CAS number 25812-30-0.
- Supplied as a 10 MG powder with recommended storage at -20°C for long-term stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.