Learn More
Medchemexpress LLC 5-[2-(dimethylamino)ethylamino]-2-methyl-1,3-benzothiazole-4,7-dione | 477603-18-2 | 95.5% | 50 MG

Supplier: Medchemexpress LLC HY12106550MG
BN-82685 (IRC-083065) is a quinone-based CDC25 phosphatase inhibitor used in preclinical cancer research. It shows low-nanomolar to sub-micromolar activity against CDC25 family members and is supplied as a brown-to-red solid for in vitro and in vivo studies.
- CAS number: 477603-18-2.
- IUPAC name: 5-[2-(dimethylamino)ethylamino]-2-methyl-1,3-benzothiazole-4,7-dione.
- Molecular formula: C12H15N3O2S.
- Molecular weight: 265.33 g/mol.
- Purity: 99.1% (manufacturer reported).
- Appearance: brown to red solid.
- Solubility: soluble in DMSO; in vivo formulation possible with DMSO, PEG300, Tween-80, and saline.
- Storage: solid at 4°C protected from light; in solution store at -80°C for long-term.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.