Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-[2-(dimethylamino)ethylamino]-2-methyl-1,3-benzothiazole-4,7-dione | 477603-18-2 | 99.1% | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1210655MG

Encompass_Preferred

BN-82685 is a small-molecule CDC25 phosphatase inhibitor supplied for research use. It is intended for biochemical and cellular assays probing phosphatase activity and cell-cycle signaling; purity should be confirmed against the lot COA before use.

  • Small-molecule CDC25 phosphatase inhibitor for research use.
  • Molecular formula C12H15N3O2S and molecular weight 265.33 g/mol.
  • Available in small pack sizes suitable for screening and assay development.
  • High reported purity for research applications, verify certificate of analysis for the supplied lot.
  • Soluble in common organic solvents at useful concentrations for in vitro studies.

Catalog No. 50-002-32769


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.