Learn More
Medchemexpress LLC 5-(2-phenylpyrazolo[1,5-a]pyridin-3-yl)-2H-pyrazolo[3,4-c]pyridazin-3-amine | 865362-74-9 | 99.9% | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1227510MM 1ML
Encompass_Preferred
FR 180204 is an ATP-competitive, selective inhibitor of ERK1 and ERK2 used for in vitro research. It inhibits ERK1 and ERK2 with IC50s of 0.51 μM and 0.33 μM, respectively, and is supplied as a solid or as a 10 mM solution in DMSO.
  • ATP-competitive ERK1/2 inhibitor with low-micromolar potency.
  • Suitable for biochemical and cell-based assays.
  • Available as a solid or 10 mM solution in DMSO.
  • High purity (≈99.9%).
  • Recommended storage preserves stability under specified conditions.
Catalog No. 50-002-30142