Learn More
Medchemexpress LLC 5-(2-thienyl)-3-isoxazolyl methanol | 194491-44-6 | MFCD03086162 | 181.21 g/mol | C8H7NO2S | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW029719100MG
Encompass_Preferred
[5-(2-Thienyl)-3-isoxazolyl]methanol is a small-molecule AgrA-DNA binding inhibitor used in research on Staphylococcus aureus virulence and infection pathways. The compound has molecular formula C8H7NO2S and a molecular weight of 181.21 g/mol. It is supplied for in vitro biochemical and microbiological studies and is reported as Compound D in the literature.
  • AgrA-DNA binding inhibitor useful for studying quorum sensing and virulence regulation.
  • Applicable to Staphylococcus aureus infection research and related assays.
  • Small-molecule reagent compatible with biochemical and microbiological assays.
  • Reported in literature as Compound D for mechanistic studies.
  • Supplied in pure form suitable for research use.
Catalog No. 50-002-99338