Learn More
Medchemexpress LLC 5-[(3,5,5,8,8-pentamethyl-6,7-dihydronaphthalen-2-yl)methyl]-N-(2,4,6-trimethoxyphenyl)furan-2-carboxa | 263847-55-8 | 99.5% | 491.62 g/mol | C30H37NO5 | 25 MG

Supplier: Medchemexpress LLC HY10753425MG
AG-045572 is a small-molecule gonadotropin-releasing hormone (GnRH) receptor antagonist for research use. It binds human and rat GnRH receptors with low-nanomolar affinity (Ki = 6.0 nM human; 3.8 nM rat), is metabolized by CYP3A, and has been used in pharmacology studies to suppress testosterone.
- CAS number 263847-55-8.
- Chemical formula C30H37NO5.
- Molecular weight 491.62 g/mol.
- Purity 99.52%.
- Appearance solid.
- Low-nanomolar GnRH receptor antagonist activity.
- Metabolized by CYP3A.
- Available in milligram-scale quantities for preclinical research.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.