Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-(3-amino-1H-indazol-6-yl)-1-[(3-chlorophenyl)methyl]pyridin-2-one | 2095704-43-9 | MFCD31728203 | 98.5% | 350.80 | C19H15ClN4O | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1121135MG

Encompass_Preferred

SLV-2436 is a small-molecule, ATP-competitive inhibitor of MAP kinase-interacting kinases MNK1 and MNK2 intended for research use to probe MNK-dependent signaling. It exhibits low-nanomolar potency and is supplied as a solid or as a DMSO solution.

  • Inhibits MNK1 (IC50 10.8 nM) and MNK2 (IC50 5.4 nM).
  • High purity (≈98.5% by HPLC).
  • Molecular formula C19H15ClN4O; molecular weight 350.80 g·mol⁻¹.
  • Available as a solid and as a 10 mM DMSO solution for easy dosing.
  • Available in small research pack sizes suitable for assay work.
  • Recommended storage: powder -20°C; in solvent -80°C for long-term storage.

Catalog No. 50-002-63564


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.