Learn More
Medchemexpress LLC 5-(3-chloro-4-cyclohexylphenyl)-1-(3-methoxyphenyl)-1H-pyrazole-3-carboxylic acid | 1393814-38-4 | 99.7% | 410.89 g·mol⁻1 | C23H23ClN2O3 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10761525MG
Encompass_Preferred
TC LPA5 4 is a small-molecule antagonist of the lysophosphatidic acid receptor 5 (LPA5/GPR92) for research use. It is a non-lipid pyrazole carboxylic acid supplied as a solid with high purity, intended for in vitro and preclinical studies of LPA5 signaling and platelet aggregation.
  • High purity to support reproducible biological assays.
  • Defined chemical identity: C23H23ClN2O3; MW 410.89 g·mol⁻1.
  • Available in small mass formats for dosing flexibility.
  • Stable when protected from light; suitable for low-temperature solvent storage.
  • Applicable to LPA5 pharmacology and platelet aggregation studies.
Catalog No. 50-002-41414