Learn More
Medchemexpress LLC 5-(3-methoxy-4-((4-methoxybenzyl)oxy)benzyl)pyrimidine-2,4-diamine | 870483-87-7 | 99.9% | 366.41 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10917200MG
Encompass_Preferred
GW2580 is an orally bioavailable and selective inhibitor of c-Fms kinase, capable of completely inhibiting human cFMS kinase in vitro at 0.06 μM. It acts as a competitive inhibitor of ATP binding to the cFMS kinase and inhibits colony-stimulating-factor-1 signaling.
  • Orally bioavailable.
  • Selective c-fms kinase inhibitor.
  • Inhibits human cFMS kinase in vitro.
  • Competitively inhibits ATP binding to cFMS kinase.
  • Inhibits colony-stimulating-factor-1 signaling.
Catalog No. 50-003-46565