Learn More
Medchemexpress LLC 5-[(3S)-3-hydroxy-4-phenyl-1-buten-1-yl]1-[6-(2H-tetrazol-5R-yl)hexyl]-2-pyrrolidinone | 346673-06-1 | MFCD18384961 | >98.0% | 383.5 g/mol | C21H29N5O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1085575MG
Encompass_Preferred
CAY10598 (TCS 2510) is a potent, selective agonist of the prostaglandin E2 EP4 receptor supplied for research use only. It activates EP4 signaling and elevates intracellular cAMP, making it a useful pharmacological tool to probe EP4-mediated pathways in inflammation, metabolic disease, bone healing, and cardiovascular research. Reported properties include Ki ≈ 1.2 nM, molecular weight 383.5 g/mol, and typical purity ≥98%.
  • Selective EP4 receptor agonist.
  • Potent EP4 binding (Ki ≈ 1.2 nM).
  • High purity ≥98%.
  • Elevates intracellular cAMP in target cells.
  • Useful for inflammation and metabolic disease research.
  • Supplied as a solid for in vitro and in vivo studies; store at -20 °C.
Catalog No. 50-002-63817