Learn More
Medchemexpress LLC 5-(4-Hydroxyphenyl)-3H-1,2-dithiole-3-thione | 18274-81-2 | 98.6% | 226.35 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY10958210MG
Encompass_Preferred
ADT-OH is a hydrogen sulfide-releasing donor that induces apoptosis and upregulates FADD, inhibiting FAK/Paxillin pathways. This compound shows potential for cancer research, demonstrating significant anti-metastatic activity in cellular studies on TNBC cells and in various mouse models, with limited effect on primary tumor growth.
  • Induces apoptosis and upregulates FADD
  • Inhibits FAK/Paxillin signaling pathways
  • Shows anti-metastatic activity on malignant melanoma cells
  • Enhances antitumor activity of celecoxib in colorectal cancer cells
  • Promotes self-renewal and antiapoptosis ability of cultured NPCs
Catalog No. 50-003-44765