Learn More
Medchemexpress LLC 5-(4-Phenylbutoxy)psoralen | 724709-68-6 | 99.7% | 334.37 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY1085831MG
Encompass_Preferred
Psora-4 is a potent and selective inhibitor of Kv1.3 (voltage-gated potassium channels) with an EC50 of 3 nM. It exhibits immunosuppressive activity and, in vitro, inhibits the proliferation of human and rat myelin-specific effector memory T cells.
  • Potent and selective inhibitor of Kv1.3.
  • EC50 of 3 nM for Kv1.3.
  • Immunosuppressive activity.
  • Inhibits proliferation of human and rat myelin-specific effector memory T cells in vitro.
Catalog No. 50-003-46276