Learn More
Medchemexpress LLC 5,5,5-trifluoro-DL-leucine | 2792-72-5 | MFCD00039528 | 98.0% | 185.14 | C6H10F3NO2 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW037549
Encompass_Preferred
5,5,5-Trifluoro-DL-leucine is a trifluoromethyl-substituted analogue of the amino acid leucine provided as a solid research reagent. It is used as a fluorinated building block in biochemical, peptide, and medicinal chemistry studies to probe metabolic stability and alter physicochemical properties. Typical purity is 98.0% and it is soluble in water when acidified.
Catalog No. 50-003-03835