Learn More
Medchemexpress LLC (5,5-dioxo-4H-thieno[3,2-c]thiochromen-2-yl)-[4-(4-methoxyphenyl)piperazin-1-yl]methanone | 890819-86-0 | MFCD14808090 | >98.0% | 454.56 g/mol | C23H22N2O4S2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY100737100MG
Encompass_Preferred
ML349 is a selective, reversible inhibitor of acyl protein thioesterase 2 (APT2)/lysophospholipase 2 (LYPLA2) used in biochemical and cell biology research to probe protein depalmitoylation. It shows reported nanomolar potency and is provided as a solid research reagent for laboratory studies.
  • Selective APT2/LYPLA2 inhibition with low-nanomolar potency.
  • Reversible inhibition suitable for mechanistic studies.
  • Enables investigation of protein palmitoylation and signaling pathways.
  • Supplied as a stable powder for low-temperature storage.
  • Characterized with CAS, formula, and molecular weight for reproducibility.
Catalog No. 50-002-12101