Learn More
Medchemexpress LLC 5-[(5-nitro-1,3-thiazol-2-yl)sulfanyl]-1,3,4-thiadiazol-2-amine | 40045-50-9 | MFCD01927019 | 98.9% | 261.3 g/mol | C5H3N5O2S3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10759710MG
Encompass_Preferred
Halicin (SU3327) is a small-molecule c-Jun N-terminal kinase (JNK) inhibitor provided for laboratory research use. It demonstrates activity against JNK1 (IC50 = 0.7 μM) and disrupts the JNK-JIP protein-protein interaction (IC50 = 239 nM); available in multiple solid and solution formats with reported high purity.
Catalog No. 50-002-40708