Learn More
Chemglass Life Sciences 5.5in O-Ring Flat Flange, Tube ID: 130mm, Tube OD: 140mm, O-Ring Size 359
SDP
Supplier:  Chemglass Life Sciences CG13805
Encompass_Preferred
5.5in O-Ring Flat Flange, Tube ID: 130mm, Tube OD: 140mm, O-Ring Size 359. Similar to CG-135 flange but with the mating end of the flange having a precision formed o-ring groove instead of being ground. Joint provides a grease-free vacuum tight seal when used with the CG-139 clamp. O-ring is included with each flange and can be ordered separately, see CG-305.
Catalog No. CG13805