Learn More
Medchemexpress LLC 5,6-dichlorobenzimidazole riboside | 53-85-0 | 99.9% | C12H12Cl2N2O4 | 10MG
SDP
Supplier:  Medchemexpress LLC HY1439210MG
Encompass_Preferred
5,6-Dichlorobenzimidazole riboside (DRB) is a chlorinated benzimidazole nucleoside analog used in molecular and cell biology research as an inhibitor of RNA polymerase II and certain cyclin-dependent kinases. It is provided as a high-purity solid or DMSO solution for use in biochemical assays, transcriptional studies, and apoptosis research.
  • Inhibits RNA polymerase II transcription in cell and biochemical assays.
  • Inhibits certain cyclin-dependent kinases for kinase studies.
  • High purity suitable for sensitive biochemical experiments.
  • Available as solid and as DMSO solution for assay flexibility.
  • Used to study transcriptional regulation and apoptosis mechanisms.
Catalog No. 50-002-58292