Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5,6-Dihydro-5-methyluracil | 696-04-8 | 50 MG
SDP

Supplier:  Medchemexpress LLC HYN678750MG

Encompass_Preferred

5,6-Dihydro-5-methyluracil (Dihydrothymine) is an intermediate breakdown product of thymine, found in animals or plants. It can be toxic at abnormally high levels.

  • Endogenous metabolite
  • Molecular weight: 128.13
  • Formula: C5H8N2O2
  • Appearance: Solid
  • Color: White to off-white
  • Initial source: Endogenous metabolite, human gut microbiota metabolites
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, sealed storage, away from moisture
  • In solvent storage: -80°C, 6 months; -20°C, 1 month (sealed storage, away from moisture)
  • Purity: 99.88%
  • Chemical structure: O=C1NC(C(C)CN1)=O
  • Structure classification: Others

Catalog No. 50-004-17918


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.