Learn More
Medchemexpress LLC 5,6-Dihydro-5-methyluracil | 696-04-8 | MFCD00023159 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN678710MM/1ML
Encompass_Preferred
5,6-Dihydro-5-methyluracil, also known as Dihydrothymine, is an intermediate breakdown product of thymine. It can be found in animals or plants and may be toxic at abnormally high levels.
  • Intermediate breakdown product of thymine
  • Found in animals or plants
  • Supplied as a 10 mM solution in DMSO
Catalog No. 50-004-20201