Learn More
Medchemexpress LLC 5,6-diphenyl-N-((tetrahydrofuran-2-yl)methyl)furo[2,3-d]pyrimidin-4-amine | 873305-35-2 | MFCD22417088 | 100.0% | 371.43 g·mol^-1 | C23H21N3O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1529010MG
Encompass_Preferred
AIM-100 is a small-molecule inhibitor of the non-receptor tyrosine kinase Ack1 (TNK2), used in biochemical and cellular research to probe Ack1-dependent signaling. It exhibits low-nanomolar potency and is supplied as a high-purity solid suitable for formulation in DMSO for in vitro assays and cell-based studies.
  • Potent Ack1 (TNK2) inhibitor with low-nanomolar activity.
  • High chemical purity suitable for biochemical and cell-based assays.
  • Soluble in DMSO for preparation of concentrated stock solutions.
  • Stable solid form for routine storage and handling.
  • Commonly used for pathway interrogation, mechanism-of-action studies, and target validation.
  • Provided with standard identifiers (CAS, formula, molecular weight) for traceability.
Catalog No. 50-002-86198