Learn More
Medchemexpress LLC 5,6-trans-Travoprost | 1563176-59-9 | >95% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0584A1MG
Encompass_Preferred
5,6-trans-Travoprost is an isomer of Travoprost and can be utilized as an experimental control. Travoprost (Fluprostenol isopropyl ester), an isopropyl ester proagent, is a high-affinity, selective FP prostaglandin full receptor agonist. It demonstrates ocular hypotensive efficacy and holds potential for treating glaucoma and ocular hypertension.
  • Isomer of Travoprost
  • Can be used as an experimental control
  • High-affinity, selective FP prostaglandin full receptor agonist
  • Demonstrates ocular hypotensive efficacy
  • Potential for treating glaucoma and ocular hypertension
Catalog No. 50-004-05908