Learn More
Medchemexpress LLC 5,6-trans-Vitamin D2 | 51744-66-2 | 97.0% | C28H44O | 5 MG
SDP
Supplier:  Medchemexpress LLC HY76542A5MG
Encompass_Preferred
5,6-trans-Vitamin D2 is an isomer of Vitamin D2, intended for research use only and not for patient administration.
  • Solid storage at -20°C, protected from light and stored under nitrogen.
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light and stored under nitrogen.
  • Appears as an off-white to light yellow solid.
Catalog No. 50-004-32275